Learn More
Grainger Metal Hose 4 ID x 18 ft.L
Catalog No. 502768498
Encompass_Preferred
The Hose Master Interlocked Metal Hose is a 4-inch diameter metal hose with a working pressure of 300 PSI. It has interlocking annular corrugations for flexibility and is made from 304 stainless steel with a mirror finish. This hose meets SAE 100R2 standards and has a 4-bolt split flange on each end. It can handle temperatures from -40 to 1000 degrees Fahrenheit. The metal hose is 30 inches long and weighs approximately 15 pounds. It is designed for conveying air, water, oil and other fluids in high pressure industrial applications.
Catalog No. 50-276-8498 Supplier Grainger Supplier No. 16T857
The Hose Master Interlocked Metal Hose is a 4-inch diameter metal hose with a working pressure of 300 PSI. It has interlocking annular corrugations for flexibility and is made from 304 stainless steel with a mirror finish. This hose meets SAE 100R2 standards and has a 4-bolt split flange on each end. It can handle temperatures from -40 to 1000 degrees Fahrenheit. The metal hose is 30 inches long and weighs approximately 15 pounds. It is designed for conveying air, water, oil and other fluids in high pressure industrial applications.