Learn More
Grainger Metal Hose 4 ID x 6 ft.L
Catalog No. 502767701
Encompass_Preferred
The Hose Master Flexible Metal Hose Assembly is a stainless steel flexible metal hose with a 4" diameter and a 12" length. It has 150 psi working pressure and 300 psi burst pressure. The hose has a stainless steel braid reinforcement and can withstand temperatures from -40°F to 1000°F. It has male NPT thread connections on both ends and includes 360° swivel flange fittings. The flexible metal hose is designed for connecting rigid piping systems where vibration dampening, noise reduction, and movement absorption is needed. It is made in the USA.
Catalog No. 50-276-7701 Supplier Grainger Supplier No. 16T799
The Hose Master Flexible Metal Hose Assembly is a stainless steel flexible metal hose with a 4" diameter and a 12" length. It has 150 psi working pressure and 300 psi burst pressure. The hose has a stainless steel braid reinforcement and can withstand temperatures from -40°F to 1000°F. It has male NPT thread connections on both ends and includes 360° swivel flange fittings. The flexible metal hose is designed for connecting rigid piping systems where vibration dampening, noise reduction, and movement absorption is needed. It is made in the USA.