Learn More
Grainger Metal Hose 4 ID x 8 ft.L
Catalog No. 502768496
Encompass_Preferred
The Grainger Approved Metal Hose is a flexible metal hose that connects air, water, oil, or exhaust systems. It has a 304 stainless steel inner hose covered by a stainless steel wire braid exterior. The metal hose is available in sizes ranging from 1/4 inch to 2 inches in diameter, with lengths from 10 feet to 50 feet. It can withstand temperatures from -425°F to 1200°F and pressures up to 500 psi. The metal hose is designed for automotive, industrial, and commercial applications that require a durable, high temperature and pressure hose.
Catalog No. 50-276-8496 Supplier Grainger Supplier No. 16T852
The Grainger Approved Metal Hose is a flexible metal hose that connects air, water, oil, or exhaust systems. It has a 304 stainless steel inner hose covered by a stainless steel wire braid exterior. The metal hose is available in sizes ranging from 1/4 inch to 2 inches in diameter, with lengths from 10 feet to 50 feet. It can withstand temperatures from -425°F to 1200°F and pressures up to 500 psi. The metal hose is designed for automotive, industrial, and commercial applications that require a durable, high temperature and pressure hose.