Learn More
Grainger Metal Hose 5 ID x 10 ft.L
Catalog No. 502768409
Encompass_Preferred
The Grainger 10 ft Corrugated Metal Hose is a flexible metal hose made of stainless steel or galvanized steel. It has a corrugated design that allows it to expand and contract. The hose has a 1-1/2" inside diameter and is rated for pressures up to 150 psi. It can be used for exhaust, fume removal, and ventilation applications. The corrugated design allows for easy routing and installation. The stainless steel version provides corrosion resistance. The galvanized steel version is more economical. This durable metal hose is ideal for industrial applications requiring flexibility.
Catalog No. 50-276-8409 Supplier Grainger Supplier No. 16T862
The Grainger 10 ft Corrugated Metal Hose is a flexible metal hose made of stainless steel or galvanized steel. It has a corrugated design that allows it to expand and contract. The hose has a 1-1/2" inside diameter and is rated for pressures up to 150 psi. It can be used for exhaust, fume removal, and ventilation applications. The corrugated design allows for easy routing and installation. The stainless steel version provides corrosion resistance. The galvanized steel version is more economical. This durable metal hose is ideal for industrial applications requiring flexibility.