Learn More
Grainger Metal Hose 5 ID x 12 ft.L
Catalog No. 502768319
Encompass_Preferred
The Grainger Approved Metal Hose is a flexible metal hose that connects air, water, oil, or exhaust systems. It has a 304 stainless steel inner hose covered by a stainless steel wire braid exterior. The hose has a temperature range of -65°F to 1000°F, a 300 PSI working pressure, and 1/4" NPT male threaded ends. The metal hose is available in sizes ranging from 1/4" to 2" diameter and lengths from 12" to 20'. It meets SAE J517 specifications. The metal hose provides durability while absorbing vibration and compensating for misalignment.
Catalog No. 50-276-8319 Supplier Grainger Supplier No. 16T863
The Grainger Approved Metal Hose is a flexible metal hose that connects air, water, oil, or exhaust systems. It has a 304 stainless steel inner hose covered by a stainless steel wire braid exterior. The hose has a temperature range of -65°F to 1000°F, a 300 PSI working pressure, and 1/4" NPT male threaded ends. The metal hose is available in sizes ranging from 1/4" to 2" diameter and lengths from 12" to 20'. It meets SAE J517 specifications. The metal hose provides durability while absorbing vibration and compensating for misalignment.