Learn More
Grainger Metal Hose 5 ID x 14 ft.L
Catalog No. 502768656
Encompass_Preferred
The Grainger 16T864 is a Dayton 1/3 HP 115/230V AC electric motor with a totally enclosed fan cooled enclosure. It has a 1,725 RPM base speed and operates on single phase power. The motor frame is 56C with a 48Y frame size. It has a 0.8 service factor and is CSA certified. The motor dimensions are 6-1/2" diameter x 7-7/16" long. It can be used for a wide variety of industrial and commercial applications requiring a small fractional horsepower AC electric motor.
Catalog No. 50-276-8656 Supplier Grainger Supplier No. 16T864
The Grainger 16T864 is a Dayton 1/3 HP 115/230V AC electric motor with a totally enclosed fan cooled enclosure. It has a 1,725 RPM base speed and operates on single phase power. The motor frame is 56C with a 48Y frame size. It has a 0.8 service factor and is CSA certified. The motor dimensions are 6-1/2" diameter x 7-7/16" long. It can be used for a wide variety of industrial and commercial applications requiring a small fractional horsepower AC electric motor.