Learn More
Grainger Metal Hose 5 ID x 14 ft.L
Catalog No. 502766647
Encompass_Preferred
The Grainger 16T812 is a Dayton 1/3 HP 115/230V AC electric motor. It is a single phase, open drip proof motor with a 1/3 horsepower rating and a 1725 RPM base speed. The motor operates on either 115V or 230V power. It features a 48 frame size with a 5/8" diameter shaft and CCW rotation when viewed from the shaft end. The motor dimensions are 6-1/2" long by 4-3/8" wide by 5-1/4" high. It weighs approximately 13 pounds and has a 1.1 service factor. The 16T812 is thermally protected for overload conditions.
Catalog No. 50-276-6647 Supplier Grainger Supplier No. 16T812
The Grainger 16T812 is a Dayton 1/3 HP 115/230V AC electric motor. It is a single phase, open drip proof motor with a 1/3 horsepower rating and a 1725 RPM base speed. The motor operates on either 115V or 230V power. It features a 48 frame size with a 5/8" diameter shaft and CCW rotation when viewed from the shaft end. The motor dimensions are 6-1/2" long by 4-3/8" wide by 5-1/4" high. It weighs approximately 13 pounds and has a 1.1 service factor. The 16T812 is thermally protected for overload conditions.