Learn More
Grainger Metal Hose 5 ID x 16 ft.L
Catalog No. 502765034
Encompass_Preferred
The Hose Master Interlocked Metal Hose is a 5-inch diameter metal hose with a working pressure of 300 PSI. It has interlocked annular corrugations for flexibility and is made from 304 stainless steel with a mirror finish. This hose meets SAE 100R2 standards and has a temperature range of -40°F to 1000°F. It comes in a standard length of 25 feet and has male NPT threaded ends. The interlocked design allows tight bending radii without kinking. This durable metal hose is ideal for conveying air, water, oil and other fluids in high pressure industrial applications.
Catalog No. 50-276-5034 Supplier Grainger Supplier No. 16T813
The Hose Master Interlocked Metal Hose is a 5-inch diameter metal hose with a working pressure of 300 PSI. It has interlocked annular corrugations for flexibility and is made from 304 stainless steel with a mirror finish. This hose meets SAE 100R2 standards and has a temperature range of -40°F to 1000°F. It comes in a standard length of 25 feet and has male NPT threaded ends. The interlocked design allows tight bending radii without kinking. This durable metal hose is ideal for conveying air, water, oil and other fluids in high pressure industrial applications.