Learn More
Grainger Metal Hose 5 ID x 18 ft.L
Catalog No. 502766464
Encompass_Preferred
The Hose Master Interlocked Metal Hose is a 5-inch diameter metal hose with a working pressure of 300 PSI. It has interlocked annular corrugations for flexibility and crush resistance. The hose is made from 304 stainless steel with a mirror-polished finish. It has a 5000 PSI burst pressure and can handle temperatures from -425 to 1000 degrees Fahrenheit. The metal hose features 316 stainless steel flanges on each end and is 20 feet in length. This durable metal hose is ideal for conveying liquids, gases, powders, and bulk materials.
Catalog No. 50-276-6464 Supplier Grainger Supplier No. 16T814
The Hose Master Interlocked Metal Hose is a 5-inch diameter metal hose with a working pressure of 300 PSI. It has interlocked annular corrugations for flexibility and crush resistance. The hose is made from 304 stainless steel with a mirror-polished finish. It has a 5000 PSI burst pressure and can handle temperatures from -425 to 1000 degrees Fahrenheit. The metal hose features 316 stainless steel flanges on each end and is 20 feet in length. This durable metal hose is ideal for conveying liquids, gases, powders, and bulk materials.