Learn More
Grainger Metal Hose 5 ID x 20 ft.L
Catalog No. 502766615
Encompass_Preferred
The Hose Master Interlocked Metal Hose is a 5-inch diameter metal hose with a working pressure of 300 PSI. It has interlocked annular corrugations for flexibility and is made from 304 stainless steel with a mirror finish. This hose meets SAE J517 standards and has a temperature range of -40°F to 1000°F. It comes in a standard length of 25 feet and has male NPT thread connections on both ends. The interlocked design allows it to be bent to a 4-inch minimum bend radius. This durable stainless steel hose is ideal for conveying air, water, oil and other fluids in high pressure, high temperature applications.
Catalog No. 50-276-6615 Supplier Grainger Supplier No. 16T815
The Hose Master Interlocked Metal Hose is a 5-inch diameter metal hose with a working pressure of 300 PSI. It has interlocked annular corrugations for flexibility and is made from 304 stainless steel with a mirror finish. This hose meets SAE J517 standards and has a temperature range of -40°F to 1000°F. It comes in a standard length of 25 feet and has male NPT thread connections on both ends. The interlocked design allows it to be bent to a 4-inch minimum bend radius. This durable stainless steel hose is ideal for conveying air, water, oil and other fluids in high pressure, high temperature applications.