Learn More
Grainger Metal Hose 5 ID x 25 ft.L
Catalog No. 502768578
Encompass_Preferred
The Hose Master Interlocked Metal Hose is a 5-inch diameter metal hose with a working pressure of 300 PSI. It has interlocking annular corrugations for flexibility and is made from 304 stainless steel with a smooth bore. This hose meets SAE 100R2 standards and has a temperature range of -40°F to 1000°F. It comes in a 25 ft length with male NPT threaded ends and is ideal for conveying air, water, oil and other fluids in industrial applications. The corrugated stainless steel construction makes it very durable while still allowing flexibility.
Catalog No. 50-276-8578 Supplier Grainger Supplier No. 16T868
The Hose Master Interlocked Metal Hose is a 5-inch diameter metal hose with a working pressure of 300 PSI. It has interlocking annular corrugations for flexibility and is made from 304 stainless steel with a smooth bore. This hose meets SAE 100R2 standards and has a temperature range of -40°F to 1000°F. It comes in a 25 ft length with male NPT threaded ends and is ideal for conveying air, water, oil and other fluids in industrial applications. The corrugated stainless steel construction makes it very durable while still allowing flexibility.