Learn More
Grainger Metal Hose 5 ID x 6 ft.L
Catalog No. 502765102
Encompass_Preferred
The Hose Master Interlocked Metal Hose is a 5-inch diameter metal hose with interlocked construction. It is made from 304 stainless steel with a maximum working pressure of 300 PSI and temperature range of -40°F to 1000°F. This flexible metal hose has a corrugated design and is available in 25-foot lengths. It can be used for conveying air, water, oil and other fluids in industrial applications. The interlocked construction allows it to be bent and shaped while maintaining strength and leak resistance.
Catalog No. 50-276-5102 Supplier Grainger Supplier No. 16T860
The Hose Master Interlocked Metal Hose is a 5-inch diameter metal hose with interlocked construction. It is made from 304 stainless steel with a maximum working pressure of 300 PSI and temperature range of -40°F to 1000°F. This flexible metal hose has a corrugated design and is available in 25-foot lengths. It can be used for conveying air, water, oil and other fluids in industrial applications. The interlocked construction allows it to be bent and shaped while maintaining strength and leak resistance.