Learn More
Grainger Metal Hose 6 ID x 10 ft.L
Catalog No. 502766479
Encompass_Preferred
The Hose Master Interlocked Metal Hose is a 6 inch diameter metal hose with a working pressure of 300 PSI. It has interlocking annular corrugations and is made from 304 stainless steel with a smooth bore. The hose has a temperature range of -40°F to 1000°F. It has a zinc electroplated finish and is 30 feet in length. The hose has a 6 inch NPT male end connection on one end and a 6 inch NPT female swivel connection on the other end. It can be used for conveying air, water, oil and other fluids. The hose is crush and kink resistant.
Catalog No. 50-276-6479 Supplier Grainger Supplier No. 16T818
The Hose Master Interlocked Metal Hose is a 6 inch diameter metal hose with a working pressure of 300 PSI. It has interlocking annular corrugations and is made from 304 stainless steel with a smooth bore. The hose has a temperature range of -40°F to 1000°F. It has a zinc electroplated finish and is 30 feet in length. The hose has a 6 inch NPT male end connection on one end and a 6 inch NPT female swivel connection on the other end. It can be used for conveying air, water, oil and other fluids. The hose is crush and kink resistant.