Learn More
Grainger Metal Hose 6 ID x 12 ft.L
Catalog No. 502766466
Encompass_Preferred
The Hose Master Interlocked Metal Hose is a 6-inch diameter metal hose with a working pressure of 300 PSI. It has interlocked annular corrugations for flexibility and is made from 304 stainless steel with a mirror finish. This hose meets military specification MIL-H-24606B and features reusable fittings that allow for repeated assembly and disassembly. With a minimum bend radius of 12 inches, this durable hose is ideal for applications involving petroleum products, chemicals, gases, water, steam or air.
Catalog No. 50-276-6466 Supplier Grainger Supplier No. 16T819
The Hose Master Interlocked Metal Hose is a 6-inch diameter metal hose with a working pressure of 300 PSI. It has interlocked annular corrugations for flexibility and is made from 304 stainless steel with a mirror finish. This hose meets military specification MIL-H-24606B and features reusable fittings that allow for repeated assembly and disassembly. With a minimum bend radius of 12 inches, this durable hose is ideal for applications involving petroleum products, chemicals, gases, water, steam or air.