Learn More
Grainger Metal Hose 6 ID x 16 ft.L
Catalog No. 502763917
Encompass_Preferred
The Grainger 16" Stainless Steel Corrugated Metal Hose has a 1" ID and is made of 304 stainless steel. It has a working pressure of 150 PSI and temperature range of -425°F to 1000°F. The corrugated design allows flexibility. Common applications include connecting rigid piping systems that require vibration absorption or motion compensation. This high quality stainless hose meets SAE J517 standard. With proper fittings, it provides a durable, flexible metal hose solution.
Catalog No. 50-276-3917 Supplier Grainger Supplier No. 16T821
The Grainger 16" Stainless Steel Corrugated Metal Hose has a 1" ID and is made of 304 stainless steel. It has a working pressure of 150 PSI and temperature range of -425°F to 1000°F. The corrugated design allows flexibility. Common applications include connecting rigid piping systems that require vibration absorption or motion compensation. This high quality stainless hose meets SAE J517 standard. With proper fittings, it provides a durable, flexible metal hose solution.