Learn More
Grainger Metal Hose 6 ID x 20 ft.L
Catalog No. 502765770
Encompass_Preferred
The Hose Master Interlocked Metal Hose is a 6 inch diameter metal hose with a working pressure of 300 PSI. It is made from 304 stainless steel with a smooth bore interior and is interlocked for flexibility. This metal hose has a temperature range of -20 to 650 degrees Fahrenheit. It has zinc-plated carbon steel fittings and is 30 feet in length. The hose weighs approximately 85 pounds and has a 4 inch minimum bend radius. It meets SAE J517 specifications. This durable stainless steel metal hose is ideal for conveying air, water, oil and other fluids in high pressure industrial applications.
Catalog No. 50-276-5770 Supplier Grainger Supplier No. 16T876
The Hose Master Interlocked Metal Hose is a 6 inch diameter metal hose with a working pressure of 300 PSI. It is made from 304 stainless steel with a smooth bore interior and is interlocked for flexibility. This metal hose has a temperature range of -20 to 650 degrees Fahrenheit. It has zinc-plated carbon steel fittings and is 30 feet in length. The hose weighs approximately 85 pounds and has a 4 inch minimum bend radius. It meets SAE J517 specifications. This durable stainless steel metal hose is ideal for conveying air, water, oil and other fluids in high pressure industrial applications.