Learn More
ERA WATERS CORP METAL IN WATER BLIND
SDP
Catalog No. NC9579043
Encompass
Metal in Water Blind,Specifically designed for drinking levels. Use for ICP and AA methods.
Catalog No. NC9579043 Supplier ERA WATERS CORP Supplier No. 590
Metal in Water Blind,Specifically designed for drinking levels. Use for ICP and AA methods.