Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals Metal Lysing Tube with Grinding Cylinder

Catalog No. MP01001771 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Capacity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
MP01001771 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP01001771 Supplier MP Biomedicals Supplier No. 116957002
Add to Cart
Add to Cart

Titanium Lysing Matrix tubes are ideal for grinding, lysing, and homogenizing your most resistant samples.

Titanium Lysing Matrix tubes are made from solid, high-quality titanium blocks, offering exceptional strength and durability compared to plastic tubes. The angled tube bottom provides a superior impact surface for effective grinding.

The titanium screw cap ensures a tight, leak-proof seal without needing plastic or rubber caps. A Teflon O-ring prevents leaks and can be cleaned, sterilized, or replaced between uses. Straight grooves on the cap improve grip for easy opening and closing.

Application Notes:

  • Dry grinding of very tough or hard samples in which generated heat can damage plastic tubes
  • Cryogenic dry grinding, where severe cold temperatures (dry ice or liquid nitrogen) can damage plastic tubes
  • Milling or grinding non-biological samples where plastic contamination is of concern
  • Sample processing with solvents or chemicals that are incompatible with plastics
TRUSTED_SUSTAINABILITY

Specifications

Capacity 50 mL
Format Tube
Isolation Technology Alkaline Lysis
Includes Grinding Cylinder
Product Type Metal Lysing Tube
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.