Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Metal MidiPrep™ Adapter for Tube Holder on FastPrep-24™

Catalog No. MP116002544 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Blade Material:
Stainless Steel
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Blade Material
MP116002544 Stainless Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116002544 Supplier MP Biomedicals™ Supplier No. 116002544
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This adapter holds lysing matrix tubes. This is the strongest FastPrep adapter, intended for the most difficult and long-lasting lysis applications.

  • Adapter for FastPrep-24 5G and Classic
  • Holds 18 x 5 mL tubes
  • Grind or homogenize most difficult samples
  • Can resist high temperatures up to 450°C
TRUSTED_SUSTAINABILITY

Specifications

Product Type Adapter
Blade Material Stainless Steel
Closure Color Blue
For Use With (Equipment) 18 x 5 mL Tube Holder on FastPrep-24

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.