Learn More
Mitegen Metal Pin Lid For Use With Nanosoft Grid Boxes
SDP
Catalog No. 5000494994
Encompass
The Metal Pin Lid performs the same as traditional plastic pin lids, however the higher quality aluminum material is more robust and more importantly reduces the likelihood of static electricity building up on the lid, as can be observed with plastic pin lids. Designed for use with Nanosoft pin lid grid boxes. The metal lid can more easily dissipate charge, reducing the likelihood of losing unclipped grids to your lid during sample preparation and handling. For ease of use, try the Pin Lid Manipulator Tool SKU M-CEM-NS-PLT01.
Catalog No. 50-004-94994 Supplier Mitegen Supplier No. MNSMPL1
The Metal Pin Lid performs the same as traditional plastic pin lids, however the higher quality aluminum material is more robust and more importantly reduces the likelihood of static electricity building up on the lid, as can be observed with plastic pin lids. Designed for use with Nanosoft pin lid grid boxes. The metal lid can more easily dissipate charge, reducing the likelihood of losing unclipped grids to your lid during sample preparation and handling. For ease of use, try the Pin Lid Manipulator Tool SKU M-CEM-NS-PLT01.