Learn More
DWK Life Sciences KIMBLE™ Metal Plate for Desiccator, 150 mm OD
SDP
Catalog No. 12141443 Shop All DWK Life Sciences Products
Change view
Click to view available options
For Use With (Equipment):
31200-150 and 31050-150
Catalog No. For Use With (Equipment)
12-141-443 31200-150 and 31050-150
1 options
Catalog No. 12-141-443 Supplier DWK Life Sciences Supplier No. 31251150
Add to Cart
Add to Cart
KIMBLE™ Metal Plate for Desiccator, 150 mm OD is a replacement metal plate designed for use with KIMBLE™ soda-lime vacuum desiccators with collar, ensuring a secure and reliable seal for your desiccation needs.
  • Replacement metal desiccator plate for Kimble soda-lime vacuum desiccators with collar
  • 150 mm outer diameter (OD) for compatible desiccator models
  • Metal construction for durability and long-term use
  • Designed to maintain a secure seal within the desiccator
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Outer 150 mm
Material Metal
For Use With (Equipment) 31200-150 and 31050-150
Product Type Desiccator Plate
Quantity 1