Learn More
Electron Microscopy Sciences Metal Polish Pinky
SDP
Catalog No. 50949060
Encompass_Preferred
Contains no silicone. Good for cleaning EM parts. A blue paste. Grit Distribution: Particles over 45 um diam = 0-5%; 45-10 um diam = 20-40%; 10-1.6 um diam = 10-30%; under 1.6 um diam = 40-60%.
Catalog No. 50-949-060 Supplier Electron Microscopy Sciences Supplier No. 71830
Contains no silicone. Good for cleaning EM parts. A blue paste. Grit Distribution: Particles over 45 um diam = 0-5%; 45-10 um diam = 20-40%; 10-1.6 um diam = 10-30%; under 1.6 um diam = 40-60%.