Learn More
Petroleum Analyzer Corporation Temperature sensor with plug / Pt100 / Class A / Metal / to 400°C
SDP
Catalog No. NC0888196
Encompass
Temperature sensor with plug / Pt100 / Class A / Metal / to 400°C
Catalog No. NC0888196 Supplier Petroleum Analyzer Corporation Supplier No. 103734
Temperature sensor with plug / Pt100 / Class A / Metal / to 400°C