Learn More
AVANTOR MASTERFLEX (VWR LLC) CPC (Colder) LCD23004 Metal Quick-Disconnect Hose Barb Elbow Insert; Valved, 1/4"" Tube ID, 1/4"" Flow Size
Catalog No. NC1065594
Encompass
Metal quick-disconnect couplings require a fitting body and an insert with the same flow size (order separately). Withstand vacuum to 250 psi, temperatures from -40 to 180¿F, and can be used with gases or liquids.
Catalog No. NC1065594 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3140392
Metal quick-disconnect couplings require a fitting body and an insert with the same flow size (order separately). Withstand vacuum to 250 psi, temperatures from -40 to 180¿F, and can be used with gases or liquids.