Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Metal QuickFlex™ Adapter for Tubes on FastPrep-96™

Catalog No. MP116010575 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
For Use With (Equipment):
96 x 2 mL Tubes on FastPrep-96
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
MP116010575 96 x 2 mL Tubes on FastPrep-96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116010575 Supplier MP Biomedicals™ Supplier No. 116010575
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Intended for high-throughput samples and can hold 96 lysing matrix tubes.

  • Adapter for FastPrep-96™ or FastePrep-96™ Pro
  • Holds 96 x 2 mL tubes
  • Anodized aluminum metal
  • Wide temperature range resistance
TRUSTED_SUSTAINABILITY

Specifications

Product Type Adapter
Blade Material Stainless Steel
Closure Color Blue
For Use With (Equipment) 96 x 2 mL Tubes on FastPrep-96

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.