Learn More
Chemglass Life Sciences Chemglass Metal Retaining Clips for Glass Stopcock Plugs, Size #1, Pack of 3 (CG-423-11)
SDP
Catalog No. NC2388549
Encompass_Preferred
Metal retaining clips (pack of 3) designed to apply pressure against glass stopcock plugs and barrels to prevent blow-out and loose fit while minimizing freezing. Constructed from high-carbon tempered spring steel with a green corrosion-resistant, phosphate-base finish for reliable laboratory use.
  • Designed for use with glass stopcock plugs and barrels.
  • Pack quantity: 3 clips.
  • Material: high-carbon tempered spring steel.
  • Finish: green corrosion-resistant, phosphate-base.
  • Prevents plug blow-out and reduces freezing.
  • Compatible with size #1 stopcocks.
Catalog No. NC2388549 Supplier Chemglass Life Sciences Supplier No. CG42311
Metal retaining clips (pack of 3) designed to apply pressure against glass stopcock plugs and barrels to prevent blow-out and loose fit while minimizing freezing. Constructed from high-carbon tempered spring steel with a green corrosion-resistant, phosphate-base finish for reliable laboratory use.
  • Designed for use with glass stopcock plugs and barrels.
  • Pack quantity: 3 clips.
  • Material: high-carbon tempered spring steel.
  • Finish: green corrosion-resistant, phosphate-base.
  • Prevents plug blow-out and reduces freezing.
  • Compatible with size #1 stopcocks.