Learn More
Grainger Metal Shlvng Cap 350 lb 24inx84inx48in
Catalog No. 502753495
Encompass_Preferred
The Grainger 14C774 is a Westward brand 1/4-inch drive socket set. It includes 17 metric sockets ranging from 4mm to 14mm in both standard and deep configurations. The chrome vanadium sockets have a knurled design for easy hand tightening and are stamped with the size. The set comes in a molded plastic case with a hinged lid and socket rail to keep the sockets organized. This socket set is ideal for automotive and machinery applications that require metric fasteners. The lifetime warranty provides peace of mind. With quality construction and useful sizes, this Westward socket set is a good choice for metric fastener projects.
Catalog No. 50-275-3495 Supplier Grainger Supplier No. 14C774
The Grainger 14C774 is a Westward brand 1/4-inch drive socket set. It includes 17 metric sockets ranging from 4mm to 14mm in both standard and deep configurations. The chrome vanadium sockets have a knurled design for easy hand tightening and are stamped with the size. The set comes in a molded plastic case with a hinged lid and socket rail to keep the sockets organized. This socket set is ideal for automotive and machinery applications that require metric fasteners. The lifetime warranty provides peace of mind. With quality construction and useful sizes, this Westward socket set is a good choice for metric fastener projects.