Learn More
Grainger Metal Shlvng Cap 375 lb 12inx84inx48in
Catalog No. 502753407
Encompass_Preferred
The Hallowell Metal Shelving Starter is a metal shelving unit designed for industrial use. It is made of steel with a gray epoxy powder coat finish and can hold up to 800 lbs per shelf level. The overall dimensions are 36" W x 18" D x 74" H. It comes with 4 adjustable shelves that provide flexible storage options. This shelving unit requires some assembly and is ideal for warehouses, storage rooms, garages, and other commercial/industrial settings. It is a sturdy and durable shelving solution for heavy-duty storage needs.
Catalog No. 50-275-3407 Supplier Grainger Supplier No. 14C758
The Hallowell Metal Shelving Starter is a metal shelving unit designed for industrial use. It is made of steel with a gray epoxy powder coat finish and can hold up to 800 lbs per shelf level. The overall dimensions are 36" W x 18" D x 74" H. It comes with 4 adjustable shelves that provide flexible storage options. This shelving unit requires some assembly and is ideal for warehouses, storage rooms, garages, and other commercial/industrial settings. It is a sturdy and durable shelving solution for heavy-duty storage needs.