Learn More
Grainger Metal Shlvng Cap 375 lb 12inx84inx48in
Catalog No. 502749187
Encompass_Preferred
The Grainger Approved 72"H x 36"W x 18"D 5-Shelf Steel Shelving Unit is a heavy duty shelving solution made of steel with a durable powder coat finish. It features 5 adjustable shelves that can hold up to 800 lbs evenly distributed. The shelves are adjustable in 1-1/2" increments. Comes fully assembled and ready to use. Overall dimensions are 72"H x 36"W x 18"D. Weight capacity per shelf is 800 lbs if weight is evenly distributed.
Catalog No. 50-274-9187 Supplier Grainger Supplier No. 14C782
The Grainger Approved 72"H x 36"W x 18"D 5-Shelf Steel Shelving Unit is a heavy duty shelving solution made of steel with a durable powder coat finish. It features 5 adjustable shelves that can hold up to 800 lbs evenly distributed. The shelves are adjustable in 1-1/2" increments. Comes fully assembled and ready to use. Overall dimensions are 72"H x 36"W x 18"D. Weight capacity per shelf is 800 lbs if weight is evenly distributed.