Learn More
Grainger Metal Shlvng Cap 375 lb 18inx84inx48in
Catalog No. 502753540
Encompass_Preferred
The Hallowell Metal Shelving Add-On Unit is a sturdy steel shelving system that can be added onto existing Hallowell shelving units. It measures 36" W x 18" D x 72" H and features 5 adjustable shelves with 3" height increments. The unit has a weight capacity of 800 lbs per shelf and 4000 lbs total. It is made from steel with a gray epoxy powder coat finish and requires minor assembly. The add-on unit can increase storage capacity of Hallowell shelving systems.
Catalog No. 50-275-3540 Supplier Grainger Supplier No. 14C772
The Hallowell Metal Shelving Add-On Unit is a sturdy steel shelving system that can be added onto existing Hallowell shelving units. It measures 36" W x 18" D x 72" H and features 5 adjustable shelves with 3" height increments. The unit has a weight capacity of 800 lbs per shelf and 4000 lbs total. It is made from steel with a gray epoxy powder coat finish and requires minor assembly. The add-on unit can increase storage capacity of Hallowell shelving systems.