Learn More
Grainger Metal Shlvng Cap 500 lb 12inx84inx36in
Catalog No. 502750116
Encompass_Preferred
The Hallowell Metal Shelving Add-On Unit is an all-welded steel shelving unit that can be added on to existing Hallowell shelving units to expand storage capacity. It measures 36"W x 18"D x 84"H and features 5 adjustable shelves with 3" height increments. The unit has a weight capacity of 800 lbs per shelf (4000 lbs total) when properly installed. It comes with mounting hardware for securing to existing units. This add-on shelf is made of steel with a gray epoxy powder coat finish and can be used for industrial, retail, or office storage needs.
Catalog No. 50-275-0116 Supplier Grainger Supplier No. 14C787
The Hallowell Metal Shelving Add-On Unit is an all-welded steel shelving unit that can be added on to existing Hallowell shelving units to expand storage capacity. It measures 36"W x 18"D x 84"H and features 5 adjustable shelves with 3" height increments. The unit has a weight capacity of 800 lbs per shelf (4000 lbs total) when properly installed. It comes with mounting hardware for securing to existing units. This add-on shelf is made of steel with a gray epoxy powder coat finish and can be used for industrial, retail, or office storage needs.