Learn More
Grainger Metal Shlvng Cap 500 lb 12inx84inx36in
Catalog No. 502750119
Encompass_Preferred
The Grainger General Purpose Boltless Metal Shelving is a freestanding shelving unit made of steel with 4 adjustable shelves. Each shelf can hold up to 500 lbs with a total load capacity of 2000 lbs. The shelves are adjustable in 1-inch increments on the uprights using metal clips. The unit measures 36"W x 18"D x 72"H. Assembly required using provided hardware. Can be used for general warehouse storage, retail displays, office supplies and more. Shelves should be evenly loaded for best stability.
Catalog No. 50-275-0119 Supplier Grainger Supplier No. 14C775
The Grainger General Purpose Boltless Metal Shelving is a freestanding shelving unit made of steel with 4 adjustable shelves. Each shelf can hold up to 500 lbs with a total load capacity of 2000 lbs. The shelves are adjustable in 1-inch increments on the uprights using metal clips. The unit measures 36"W x 18"D x 72"H. Assembly required using provided hardware. Can be used for general warehouse storage, retail displays, office supplies and more. Shelves should be evenly loaded for best stability.