Learn More
Grainger Metal Shlvng Cap 500 lb 12inx84inx36in
Catalog No. 502753464
Encompass_Preferred
The Grainger 14C763 is a Westward brand 1/4-inch drive socket set. This 108-piece mechanic's tool set includes sockets ranging from 4mm to 14mm in both standard and deep configurations, as well as a quick release ratchet, extension bars, adapters, a spinner handle, and universal joints. The chrome vanadium steel sockets feature a matte finish and are engraved with laser etched size markings. This socket set comes packed in a lightweight blow molded case for storage and portability.
Catalog No. 50-275-3464 Supplier Grainger Supplier No. 14C763
The Grainger 14C763 is a Westward brand 1/4-inch drive socket set. This 108-piece mechanic's tool set includes sockets ranging from 4mm to 14mm in both standard and deep configurations, as well as a quick release ratchet, extension bars, adapters, a spinner handle, and universal joints. The chrome vanadium steel sockets feature a matte finish and are engraved with laser etched size markings. This socket set comes packed in a lightweight blow molded case for storage and portability.