Learn More
Grainger Metal Shlvng Cap 500 lb 12inx84inx36in
Catalog No. 502753511
Encompass_Preferred
The Grainger General Purpose Boltless Metal Shelving is a freestanding shelving unit made of steel with 4 adjustable shelves. Each shelf has a 500 lb load capacity and the overall dimensions are 36"W x 18"D x 72"H. The shelves are adjustable in 1" increments on the uprights using metal clips with no tools required for assembly. This gray epoxy painted shelving unit can support up to 2000 lbs total load capacity.
Catalog No. 50-275-3511 Supplier Grainger Supplier No. 14C776
The Grainger General Purpose Boltless Metal Shelving is a freestanding shelving unit made of steel with 4 adjustable shelves. Each shelf has a 500 lb load capacity and the overall dimensions are 36"W x 18"D x 72"H. The shelves are adjustable in 1" increments on the uprights using metal clips with no tools required for assembly. This gray epoxy painted shelving unit can support up to 2000 lbs total load capacity.