Learn More
Grainger Metal Shlvng Cap 500 lb 12inx84inx36in
Catalog No. 502749301
Encompass_Preferred
The Grainger 14C752 is a 1/2 HP, 115V, 1725 RPM, 56C frame, open drip proof, single phase, ball bearing electric motor. It has a 1/2" diameter shaft with a 1/4" keyway and counterclockwise rotation facing the shaft end. The motor is totally enclosed, fan cooled with a cast iron frame and thermally protected for overload. It can be used for general purpose applications requiring 1/2 HP.
Catalog No. 50-274-9301 Supplier Grainger Supplier No. 14C752
The Grainger 14C752 is a 1/2 HP, 115V, 1725 RPM, 56C frame, open drip proof, single phase, ball bearing electric motor. It has a 1/2" diameter shaft with a 1/4" keyway and counterclockwise rotation facing the shaft end. The motor is totally enclosed, fan cooled with a cast iron frame and thermally protected for overload. It can be used for general purpose applications requiring 1/2 HP.