Learn More
Grainger Metal Shlvng Cap 500 lb 24inx84inx36in
Catalog No. 502752222
Encompass_Preferred
The Grainger General Purpose Boltless Metal Shelving is a freestanding shelving unit made of steel with 4 adjustable shelves. Each shelf can hold up to 500 lbs with a total load capacity of 2000 lbs. The shelves are adjustable in 1 inch increments on the uprights using metal connectors. The shelves are 36"D x 48"W. The overall dimensions are 71-7/8"H x 48"W x 18"D. This gray epoxy painted shelving unit provides economical storage solutions for warehouses, stockrooms, and other commercial or industrial settings.
Catalog No. 50-275-2222 Supplier Grainger Supplier No. 14P337
The Grainger General Purpose Boltless Metal Shelving is a freestanding shelving unit made of steel with 4 adjustable shelves. Each shelf can hold up to 500 lbs with a total load capacity of 2000 lbs. The shelves are adjustable in 1 inch increments on the uprights using metal connectors. The shelves are 36"D x 48"W. The overall dimensions are 71-7/8"H x 48"W x 18"D. This gray epoxy painted shelving unit provides economical storage solutions for warehouses, stockrooms, and other commercial or industrial settings.