Learn More
Perkin Elmer US LLC Metal Spill Tray For use with NexION 1000 and 2000 ICP-MS series instruments.
Catalog No. 502104820
Encompass_Preferred
This spill tray's dimensions are 31.12 cm by 43.82 cm and 2.54 cm deep. It features rolled edges and an uncoated 19-20 gauge Aluminum build.
Catalog No. 50-210-4820 Supplier Perkin Elmer US LLC Supplier No. N8145022
This spill tray's dimensions are 31.12 cm by 43.82 cm and 2.54 cm deep. It features rolled edges and an uncoated 19-20 gauge Aluminum build.