Learn More
Alkali Scientific Metal (Stainless Steel) Histology Embedding Base Molds for Histology & Tissue Embedding, Reusable, Polished, 24x24x9mm, 10/Pk
SDP
Catalog No. 5000662225
Encompass_Preferred
These reusable stainless steel histology base molds are designed for embedding biopsy and tissue specimens during routine histological processing. Manufactured from durable, rust resistant stainless steel, the molds provide excellent thermal conductivity for efficient and uniform paraffin embedding. Extended winged sidewalls create a stable flat bottom profile that resists tipping while offering a secure gripping surface for safe and convenient handling. The polished interior surfaces allow smooth paraffin release and simplify cleaning between uses. These molds are compatible with standard embedding cassettes and are suitable for holding paraffin and tissue samples across a wide range of laboratory workflows. Available in multiple well dimensions to accommodate different specimen s and orientations, providing consistent performance in histology laboratories.
Catalog No. 50-006-62225 Supplier Alkali Scientific Supplier No. MBM93
These reusable stainless steel histology base molds are designed for embedding biopsy and tissue specimens during routine histological processing. Manufactured from durable, rust resistant stainless steel, the molds provide excellent thermal conductivity for efficient and uniform paraffin embedding. Extended winged sidewalls create a stable flat bottom profile that resists tipping while offering a secure gripping surface for safe and convenient handling. The polished interior surfaces allow smooth paraffin release and simplify cleaning between uses. These molds are compatible with standard embedding cassettes and are suitable for holding paraffin and tissue samples across a wide range of laboratory workflows. Available in multiple well dimensions to accommodate different specimen s and orientations, providing consistent performance in histology laboratories.