Learn More
Bellco Glass, Inc. Metal Stone Sparger,15L-RB For Round Bottom Omni Vessels
SDP
Catalog No. 501944918
Encompass_Preferred
Metal stone sparger 15l round bottom. MADE IN THE USA. Metal stone sparger 15l for flat or round bottom omni vessels. Sparger inserts through stainless steal headplate fitting. 4 1/2"wide x 23 1/2" long 316 l stainless steel ra 32 finish or better Stainless
Catalog No. 50-194-4918 Supplier Bellco Glass, Inc. Supplier No. 196489415
Metal stone sparger 15l round bottom. MADE IN THE USA. Metal stone sparger 15l for flat or round bottom omni vessels. Sparger inserts through stainless steal headplate fitting. 4 1/2"wide x 23 1/2" long 316 l stainless steel ra 32 finish or better Stainless