Learn More
Bellco Glass, Inc. Metal Stone Sparger,5L-FB For Flat Bottom Omni Vessels
SDP
Catalog No. 501944912
Encompass_Preferred
Metal stone sparger 5l for flat or round bottom omni vessels. Sparger inserts through stainless steal headplate fitting, 5/16" . MADE IN THE USA. 4 1/2"wide x 12" long 316 l stainless steel ra 32 finish or better Stainless steel frit
Catalog No. 50-194-4912 Supplier Bellco Glass, Inc. Supplier No. 196489058
Metal stone sparger 5l for flat or round bottom omni vessels. Sparger inserts through stainless steal headplate fitting, 5/16" . MADE IN THE USA. 4 1/2"wide x 12" long 316 l stainless steel ra 32 finish or better Stainless steel frit