Learn More
Bellco Glass, Inc. Metal Stone Sparger,8L-RB For Round Bottom Omni Vessels
SDP
Catalog No. 501944917
Encompass_Preferred
Metal stone sparger 8l for flat or round bottom omni vessels. Sparger inserts through stainless steal headplate fitting. MADE IN THE USA. 4 1/2"wide x 15 1/2" long 316 l stainless steel ra 32 finish or better Stainless steel frit
Catalog No. 50-194-4917 Supplier Bellco Glass, Inc. Supplier No. 196489408
Metal stone sparger 8l for flat or round bottom omni vessels. Sparger inserts through stainless steal headplate fitting. MADE IN THE USA. 4 1/2"wide x 15 1/2" long 316 l stainless steel ra 32 finish or better Stainless steel frit