Learn More
Enterprise Technology Solutions Metal Surface Polish, 8 oz Bottle
SDP
Catalog No. 502110998
Encompass_Preferred
Multi-purpose metal polish for a variety of metals including brass, copper, stainless steel, chrome, aluminum, pewter, and bronze.Provides a long-lasting smooth, polished surface and a brilliant shine.Gives extra shine to stainless steel appliances.Removes tarnish.Easy to use—no rinse needed.
Catalog No. 50-211-0998 Supplier Enterprise Technology Solutions Supplier No. RAC89334
Multi-purpose metal polish for a variety of metals including brass, copper, stainless steel, chrome, aluminum, pewter, and bronze.Provides a long-lasting smooth, polished surface and a brilliant shine.Gives extra shine to stainless steel appliances.Removes tarnish.Easy to use—no rinse needed.