Learn More
Julabo USA Inc Each, 1.5m triple insulated metal tubing, M16x1f fittings
SDP
Catalog No. 507534138
Encompass_Preferred
Bath Accessories, Julabo, Metal Tubing Triple insulated (-100°C…+350°C), 1.5 m Metal tubing with connector M16x1 female fittings
Catalog No. 50-753-4138 Supplier Julabo USA Inc Supplier No. 8930211
Bath Accessories, Julabo, Metal Tubing Triple insulated (-100°C…+350°C), 1.5 m Metal tubing with connector M16x1 female fittings