Learn More
Julabo USA Inc Each, 3m single insulated metal tubing, M16x1f fittings
SDP
Catalog No. 507534143
Encompass_Preferred
Bath Accessories, Julabo, Metal Tubing Single-Insulated 3m (-50°C…+200°C) with M16x1 female fittings
Catalog No. 50-753-4143 Supplier Julabo USA Inc Supplier No. 8930223
Bath Accessories, Julabo, Metal Tubing Single-Insulated 3m (-50°C…+200°C) with M16x1 female fittings