Learn More
Julabo USA Inc Each, 0.5m single insulated metal tubing, M16x1f fittings
SDP
Catalog No. 507534140
Encompass_Preferred
Bath Accessories, Julabo, Metal Tubing Single-Insulated 0.5m (-50°C…+200°C) with M16x1 female fittings
Catalog No. 50-753-4140 Supplier Julabo USA Inc Supplier No. 8930220
Bath Accessories, Julabo, Metal Tubing Single-Insulated 0.5m (-50°C…+200°C) with M16x1 female fittings