Learn More
Grainger Little Giant Utility Cart, Steel, 5 Shelves, 2,000 lb, 1 ea
Catalog No. 502785826
Encompass_Preferred
These flow-through utility carts are designed with perforated steel shelves to allow liquids, dirt, and debris to pass through, making them ideal for use in wet or dirty environments like plant nurseries and manufacturing facilities. The welded steel construction provides a 2,000 lb total load capacity, with each of the five shelves supporting up to 400 lb.
  • Five 30 in. x 60 in. perforated shelves with 1-1/2 in. 4-sided lips to prevent sliding
  • Welded 14 ga. steel construction with a powder coated finish
  • Equipped with two rigid and two swivel 6 in. mold-on rubber casters
  • Flat handle for convenient maneuvering
  • Assembled design for immediate use
Catalog No. 50-278-5826 Supplier Grainger Supplier No. 19C139
These flow-through utility carts are designed with perforated steel shelves to allow liquids, dirt, and debris to pass through, making them ideal for use in wet or dirty environments like plant nurseries and manufacturing facilities. The welded steel construction provides a 2,000 lb total load capacity, with each of the five shelves supporting up to 400 lb.
  • Five 30 in. x 60 in. perforated shelves with 1-1/2 in. 4-sided lips to prevent sliding
  • Welded 14 ga. steel construction with a powder coated finish
  • Equipped with two rigid and two swivel 6 in. mold-on rubber casters
  • Flat handle for convenient maneuvering
  • Assembled design for immediate use