Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Metal Wave Machine
GREENER_CHOICESCI_ED

Catalog No. S139437 Shop All Supertek Scientific Products
Change view
Click to view available options
Grade:
6+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S139437 6+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S139437 Supplier Supertek™ Supplier No. PH36040
Add to Cart
Add to Cart
This item is not returnable. View return policy

Demonstrate longitudinal and transverse waves using this Supertek™ Metal Wave Machine.

  • An effective way to demonstrate longitudinal and transverse waves.
  • By turning the handle, a number of eccentric discs move a number of metal rods.
  • Transverse waves are obtained by straight rods and longitudinal waves by bent rods.
  • Comes with instruction manual.

Specifications

Grade 6+
Format Kit
Languages English
Subject Physical Science - Force and Mass
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.