Learn More
Crescent Chemical Co Inc METALAXYL 100 MG
SDP
Catalog No. 5074454
Encompass_Preferred
Metalaxyl Fungicide (WS) CAS 57837-19-1 Molecular Weight - 279.3315 Storage - RT Catalog Num - DRE-C14920000 pesticide
Catalog No. 50-744-54 Supplier Crescent Chemical Co Inc Supplier No. PSC14920000
Metalaxyl Fungicide (WS) CAS 57837-19-1 Molecular Weight - 279.3315 Storage - RT Catalog Num - DRE-C14920000 pesticide