Learn More
Medchemexpress LLC Metalaxyl-13C6 | 1356199-69-3 | 98.4% | 1 MG
SDP
Catalog No. 5000408815
Encompass_Preferred
Metalaxyl-13C6 is a 13C-labeled version of Metalaxyl, a fungicide that effectively inhibits protein synthesis in fungi. It has been shown to inhibit the growth of potato blight fungal isolates. This compound is valuable as a tracer or as an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS.
  • Used as a tracer in research
  • Functions as an internal standard for quantitative analysis
  • Inhibits protein synthesis in fungi
  • Effective against potato blight fungal isolates
Catalog No. 50-004-08815 Supplier Medchemexpress LLC Supplier No. HYB0843S1MG
Metalaxyl-13C6 is a 13C-labeled version of Metalaxyl, a fungicide that effectively inhibits protein synthesis in fungi. It has been shown to inhibit the growth of potato blight fungal isolates. This compound is valuable as a tracer or as an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS.
  • Used as a tracer in research
  • Functions as an internal standard for quantitative analysis
  • Inhibits protein synthesis in fungi
  • Effective against potato blight fungal isolates