Learn More
Medchemexpress LLC Metalaxyl-13C6 | 1356199-69-3 | 98.4% | 5 MG
SDP
Catalog No. 5000408816
Encompass_Preferred
Metalaxyl-13C6 is the 13C-labeled Metalaxyl, a fungicide known for inhibiting protein synthesis in fungi. It has been shown to inhibit the growth of potato blight (P. infestans) fungal isolates from Serbian potato fields. This compound is intended for research use only and is not for patient application.
  • 13C-labeled Metalaxyl
  • Inhibits protein synthesis in fungi
  • Inhibits potato blight (P. infestans)
  • Can be used as a tracer for quantitative analysis
  • Can be used as an internal standard for quantitative analysis
Catalog No. 50-004-08816 Supplier Medchemexpress LLC Supplier No. HYB0843S5MG
Metalaxyl-13C6 is the 13C-labeled Metalaxyl, a fungicide known for inhibiting protein synthesis in fungi. It has been shown to inhibit the growth of potato blight (P. infestans) fungal isolates from Serbian potato fields. This compound is intended for research use only and is not for patient application.
  • 13C-labeled Metalaxyl
  • Inhibits protein synthesis in fungi
  • Inhibits potato blight (P. infestans)
  • Can be used as a tracer for quantitative analysis
  • Can be used as an internal standard for quantitative analysis